Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzethonium Chloride, USP, 97-103%, Spectrum™ Chemical
SDP

Catalog No. 18600888 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-888 2.5 kg Amber Glass Bottle
18-600-883 25 g Amber Glass Bottle
18-600-884 125 g Amber Glass Bottle
18-600-885 500 g Amber Glass Bottle
18-600-886 12 kg Amber Glass Bottle
18-600-887 50 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-600-888 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. BE1292.5KG
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BE129, 121-54-0, C27H42ClNO2

Spectrum™ Chemical Benzethonium Chloride, USP exhibits a broad spectrum of microbiocidal activity against bacteria, fungi, mold and viruses. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 121-54-0
Molecular Formula C27H42ClNO2
Molecular Weight (g/mol) 448.09
InChI Key UREZNYTWGJKWBI-UHFFFAOYSA-M
IUPAC Name benzyldimethyl(2-{2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy}ethyl)azanium chloride
SMILES [Cl-].CC(C)(C)CC(C)(C)C1=CC=C(OCCOCC[N+](C)(C)CC2=CC=CC=C2)C=C1

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 5%
Packaging Amber Glass Bottle
Quantity 2.5 kg
UN Number UN3261
Percent Purity 97 to 103%
Grade USP
Physical Form Crystalline Powder or Lumps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.