accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 121-75-5

IUPAC Name: 1,4-diethyl 2-{[dimethoxy(sulfanylidene)-λ⁵-phosphanyl]sulfanyl}butanedioate
Synonyms: paladin phosphothion kop-thionkypfosmalacide kill-A-mite staeubol-puder kop-thion ortho malathion calmathion malathion (substance) 马拉硫磷
Molecular Weight (g/mol): 330.35
Molecular Formula: C10H19O6PS2
InChi Key: JXSJBGJIGXNWCI-UHFFFAOYNA-N
SMILES: CCOC(=O)CC(SP(=S)(OC)OC)C(=O)OCC
Solvent or Matrix 
  • (1)
  • (1)
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results