accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOHOHOH

CAS RN 121-79-9

IUPAC Name: propyl 3,4,5-trihydroxybenzoate
Synonyms: propyl gallate N-propyl gallate propylester kyseliny gallove propyl gallate (product) nipa 49 nipagallin P progallin P propyl gallate preparation ccris 541 tenox PG
Molecular Weight (g/mol): 212.2
Molecular Formula: C10H12O5
InChi Key: ZTHYODDOHIVTJV-UHFFFAOYSA-N
SMILES: CCCOC(=O)C1=CC(O)=C(O)C(O)=C1
Molecular Weight (g/mol) 
  • (9)
  • (6)
Quantity 
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (1)
  • (4)
  • (3)
Physical Form 
  • (5)
  • (2)
Grade 
  • (1)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (10)
  • (4)
  • (1)
  • (3)
  • (4)
  • (2)
  • (6)
  • (4)
  • (9)
  • (6)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (2)
  • (3)
  • (1)
  • (4)
  • (3)
  • (5)
  • (2)
  • (1)
  • (4)