Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Propyl Gallate MP Biomedicals

Catalog No. ICN10274750 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10274750 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10274750 Supplier MP Biomedicals, Inc Supplier No. 0210274750
Add to Cart
Add to Cart

Chemical Identifiers

CAS 121-79-9
Molecular Formula C10H12O5
Molecular Weight (g/mol) 212.201
InChI Key ZTHYODDOHIVTJV-UHFFFAOYSA-N
Synonym propyl gallate, n-propyl gallate, progallin p, tenox pg, nipagallin p, gallic acid, propyl ester, nipa 49, benzoic acid, 3,4,5-trihydroxy-, propyl ester, 3,4,5-trihydroxybenzene-1-propylcarboxylate
PubChem CID 4947
ChEBI CHEBI:10607
IUPAC Name propyl 3,4,5-trihydroxybenzoate

Specifications

Quantity 50 g
Formula Weight 212.2
Chemical Name or Material N-Propyl Gallate, “Antioxidant for foods, fats, and oils.”
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.