Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Propyl Gallate, MP Biomedicals™

Catalog No. MP210274780 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210274780 100 g
ICN10274790 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP210274780 Supplier MP Biomedicals, Inc Supplier No. 0210274780
Add to Cart
Add to Cart

Off-White Powder

  • Anti-fade reagent in fluorescence microscopy to reduce photobleaching of fluorescent probes such as rhodamine and fluorescein
  • An ester formed by the condensation of gallic acid and propanol
  • This antioxidant has been added to foods containing oils and fats to prevent oxidation
  • An antioxidant that exhibits antimicrobial activity
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-79-9
Molecular Formula C10H12O5
Molecular Weight (g/mol) 212.2
MDL Number MFCD00002196
InChI Key ZTHYODDOHIVTJV-UHFFFAOYSA-N

Specifications

Melting Point 140°C to 150°C
Color Off-white
Loss on Drying 0.0026
Quantity 100 g
Beilstein 1877976
Synonym Gallic Acid, Propyl Ester, Progallin P, n-Propyl Ester of 3,4,5-Trihydroxybenzoic Acid
Solubility Information Soluble in ethanol (50mg/mL).
Physical Form Powder
Grade 0.992
RTECSNumber LW8400000
storageNote1 Store at Room Temperature (15-30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.