accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNOOONOO

CAS RN 1210004-12-8

IUPAC Name: 4-nitrophenyl 4-[(3-phenoxyphenyl)methyl]piperazine-1-carboxylate
Synonyms: 4-nitrophenyl 4-[(3-phenoxyphenyl)methyl]piperazine-1-carboxylate
Molecular Weight (g/mol): 433.46
Molecular Formula: C24H23N3O5
InChi Key: QNYRAEKLMNDRFY-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(OC(=O)N2CCN(CC3=CC(OC4=CC=CC=C4)=CC=C3)CC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results