accessibility menu, dialog, popup

UserName

Viewing profile as user:
BFFFNHOOH

CAS RN 1212021-62-9

IUPAC Name: [3-cyano-5-(trifluoromethyl)phenyl]boronic acid
Synonyms: 3-cyano-5-(trifluoromethyl)phenylboronic acid
Molecular Weight (g/mol): 214.94
Molecular Formula: C8H5BF3NO2
InChi Key: CTJAWCWPRYNDDQ-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(=CC(=C1)C(F)(F)F)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results