accessibility menu, dialog, popup

UserName

Viewing profile as user:
ZnSSNHHNSS

CAS RN 12122-67-7

IUPAC Name: zinc(2+) ({2-[(sulfanidylmethanethioyl)amino]ethyl}carbamothioyl)sulfanide
Synonyms: zineb
Molecular Weight (g/mol): 275.73
Molecular Formula: C4H6N2S4Zn
InChi Key: AMHNZOICSMBGDH-UHFFFAOYSA-L
SMILES: [Zn++].[S-]C(=S)NCCNC([S-])=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results