accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1212272-05-3

IUPAC Name: tert-butyl N-[4-(2-aminoethyl)cyclohexyl]carbamate
Synonyms: tert-butyl N-[4-(2-aminoethyl)cyclohexyl]carbamate
Molecular Weight (g/mol): 242.36
Molecular Formula: C13H26N2O2
InChi Key: SSZKBGFWNFAXNC-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1CCC(CCN)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)