CAS RN 121247-01-6
IUPAC Name:
2-(3-fluorophenyl)-2-oxoacetaldehyde hydrate
Synonyms:
2-(3-fluorophenyl)-2-oxoacetaldehyde hydrate
Molecular Weight (g/mol):
170.14
Molecular Formula:
C8H7FO3
InChi Key:
VGFHAWFFIQFSRR-UHFFFAOYSA-N
SMILES:
O.FC1=CC=CC(=C1)C(=O)C=O
1
–
2
of
2
results
1
3-Fluorophenylglyoxal hydrate, Thermo Scientific™
CAS: 121247-01-6 Molecular Formula: C8H7FO3 Molecular Weight (g/mol): 170.14 InChI Key: VGFHAWFFIQFSRR-UHFFFAOYSA-N Synonym: 3-fluorophenylglyoxal hydrate,2-3-fluorophenyl-2-oxoacetaldehyde hydrate,acmc-20an3a,3-fluorophenyl glyoxal,3-fluoro-a-oxobenzeneacetaldehyde,benzeneacetaldehyde,3-fluoro-a-oxo,2-3-fluorophenyl-2-oxoethanal, hydrate PubChem CID: 2783267 IUPAC Name: 2-(3-fluorophenyl)-2-oxoacetaldehyde;hydrate SMILES: O.FC1=CC=CC(=C1)C(=O)C=O