Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluorophenylglyoxal hydrate, Thermo Scientific™

Catalog No. MAY00104DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MAY00104DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAY00104DA Supplier Thermo Scientific Chemicals Supplier No. MAY00104DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 121247-01-6
Molecular Formula C8H7FO3
Molecular Weight (g/mol) 170.14
InChI Key VGFHAWFFIQFSRR-UHFFFAOYSA-N
Synonym 3-fluorophenylglyoxal hydrate, 2-3-fluorophenyl-2-oxoacetaldehyde hydrate, acmc-20an3a, 3-fluorophenyl glyoxal, 3-fluoro-a-oxobenzeneacetaldehyde, benzeneacetaldehyde,3-fluoro-a-oxo, 2-3-fluorophenyl-2-oxoethanal, hydrate
PubChem CID 2783267
IUPAC Name 2-(3-fluorophenyl)-2-oxoacetaldehyde;hydrate
SMILES O.FC1=CC=CC(=C1)C(=O)C=O

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 152.12
Percent Purity 97%
Chemical Name or Material 3-fluorophenylglyoxal hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.