accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1215333-48-4

IUPAC Name: 2-{4-[4-(4-carbamimidoylphenyl)piperazin-1-yl]piperidin-1-yl}acetic acid trihydrochloride
Synonyms: {4-[4-(4-carbamimidoylphenyl)piperazin-1-yl]piperidin-1-yl}acetic acid trihydrochloride
Molecular Weight (g/mol): 454.82
Molecular Formula: C18H30Cl3N5O2
InChi Key: YKRNPHOBDOUQTG-UHFFFAOYSA-N
SMILES: Cl.Cl.Cl.NC(=N)C1=CC=C(C=C1)N1CCN(CC1)C1CCN(CC(O)=O)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results