CAS RN 1215654-26-4
IUPAC Name:
5-methyl-3-(3-{4-[2-(2,2,2-trifluoroethoxy)phenyl]piperazin-1-yl}propyl)-1,2,3,4-tetrahydropyrimidine-2,4-dione hydrochloride
Synonyms:
5-methyl-3-(3-{4-[2-(2,2,2-trifluoroethoxy)phenyl]piperazin-1-yl}propyl)-1H-pyrimidine-2,4-dione hydrochloride
Molecular Weight (g/mol):
462.9
Molecular Formula:
C20H26ClF3N4O3
InChi Key:
CWVABCXVOAVUJL-UHFFFAOYSA-N
SMILES:
Cl.CC1=CNC(=O)N(CCCN2CCN(CC2)C2=CC=CC=C2OCC(F)(F)F)C1=O
1
–
1
of
1
results
1
R&D Systems™ RS 100329 hydrochloride
CAS: 1215654-26-4 Molecular Formula: C20H26ClF3N4O3