CAS RN 1215721-40-6
IUPAC Name:
2-[(3-chlorophenyl)methoxy]-6-(piperazin-1-yl)pyrazine hydrochloride
Synonyms:
2-[(3-chlorophenyl)methoxy]-6-(piperazin-1-yl)pyrazine hydrochloride
Molecular Weight (g/mol):
341.24
Molecular Formula:
C15H18Cl2N4O
InChi Key:
NMUNRTCTDLORDR-UHFFFAOYSA-N
SMILES:
Cl.ClC1=CC=CC(COC2=NC(=CN=C2)N2CCNCC2)=C1
1
–
1
of
1
results
1
R&D Systems™ CP 809101 hydrochloride
CAS: 1215721-40-6 Molecular Formula: C15H18Cl2N4O