CAS RN 1215766-76-9
IUPAC Name:
5-chloro-N4-(cyclopropylmethyl)-2-methyl-N4-propyl-N6-(2,4,6-trichlorophenyl)pyrimidine-4,6-diamine hydrochloride
Synonyms:
5-chloro-N4-(cyclopropylmethyl)-2-methyl-N4-propyl-N6-(2,4,6-trichlorophenyl)pyrimidine-4,6-diamine hydrochloride
Molecular Weight (g/mol):
470.64
Molecular Formula:
C18H21Cl5N4
InChi Key:
CPMGENCTAWBLNW-UHFFFAOYSA-N
SMILES:
Cl.CCCN(CC1CC1)C1=NC(C)=NC(NC2=C(Cl)C=C(Cl)C=C2Cl)=C1Cl
1
–
1
of
1
results
1
R&D Systems™ NBI 27914 hydrochloride
CAS: 1215766-76-9 Molecular Formula: C18H21Cl5N4