CAS RN 1215833-62-7
IUPAC Name:
methyl 4-ethyl-6,7-dimethoxy-9H-pyrido[3,4-b]indole-3-carboxylate hydrochloride
Synonyms:
DMCM hydrochloride
Molecular Weight (g/mol):
350.8
Molecular Formula:
C17H19ClN2O4
InChi Key:
FHCRBVQGMZUJKY-UHFFFAOYSA-N
SMILES:
Cl.CCC1=C(N=CC2=C1C1=C(N2)C=C(OC)C(OC)=C1)C(=O)OC
1
–
1
of
1
results
1
R&D Systems™ DMCM hydrochloride
CAS: 1215833-62-7 Molecular Formula: C17H19ClN2O4