CAS RN 1216468-02-8
IUPAC Name:
4-iodo-N-[4-methoxy-3-(4-methylpiperazin-1-yl)phenyl]benzene-1-sulfonamide hydrochloride
Synonyms:
4-iodo-N-[4-methoxy-3-(4-methylpiperazin-1-yl)phenyl]benzenesulfonamide hydrochloride
Molecular Weight (g/mol):
523.81
Molecular Formula:
C18H23ClIN3O3S
InChi Key:
BFGOZFXMAJBFGI-UHFFFAOYSA-N
SMILES:
Cl.COC1=C(C=C(NS(=O)(=O)C2=CC=C(I)C=C2)C=C1)N1CCN(C)CC1
1
–
1
of
1
results
1
R&D Systems™ SB 258585 hydrochloride
CAS: 1216468-02-8 Molecular Formula: C18H23ClIN3O3S