CAS RN 1216540-18-9
IUPAC Name:
5-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]-2,2-diphenylpentanenitrile hydrochloride
Synonyms:
5-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]-2,2-diphenylpentanenitrile hydrochloride
Molecular Weight (g/mol):
481.46
Molecular Formula:
C28H30Cl2N2O
InChi Key:
SSZWNUGWOGONQJ-UHFFFAOYSA-N
SMILES:
Cl.OC1(CCN(CCCC(C#N)(C2=CC=CC=C2)C2=CC=CC=C2)CC1)C1=CC=C(Cl)C=C1
1
–
1
of
1
results
1
R&D Systems™ BX 513 hydrochloride
CAS: 1216540-18-9 Molecular Formula: C28H30Cl2N2O