accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1216665-49-4

IUPAC Name: N2-(1H-1,3-benzodiazol-6-yl)-N4-(5-cyclopropyl-1H-pyrazol-3-yl)pyrimidine-2,4-diamine
Synonyms: N2-(3H-1,3-benzodiazol-5-yl)-N4-(5-cyclopropyl-1H-pyrazol-3-yl)pyrimidine-2,4-diamine
Molecular Weight (g/mol): 332.37
Molecular Formula: C17H16N8
InChi Key: WJNBSTLIALIIEW-UHFFFAOYSA-N
SMILES: C1CC1C1=CC(NC2=NC(NC3=CC=C4N=CNC4=C3)=NC=C2)=NN1
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results