CAS RN 1216720-69-2
IUPAC Name:
N3-cyclopropyl-7-{[4-(propan-2-yl)phenyl]methyl}-7H-pyrrolo[3,2-f]quinazoline-1,3-diamine dihydrochloride
Synonyms:
N3-cyclopropyl-7-[(4-isopropylphenyl)methyl]pyrrolo[3,2-f]quinazoline-1,3-diamine dihydrochloride
Molecular Weight (g/mol):
444.4
Molecular Formula:
C23H27Cl2N5
InChi Key:
NNJTXSQXGHYXAJ-UHFFFAOYSA-N
SMILES:
Cl.Cl.CC(C)C1=CC=C(CN2C=CC3=C2C=CC2=NC(NC4CC4)=NC(N)=C32)C=C1
1
–
1
of
1
results
1
R&D Systems™ SCH 79797 dihydrochloride
CAS: 1216720-69-2 Molecular Formula: C23H27Cl2N5