CAS RN 1216905-73-5
IUPAC Name:
2-hydroxy-5-{2-[(2-hydroxy-3-{4-[1-methyl-4-(trifluoromethyl)-1H-imidazol-2-yl]phenoxy}propyl)amino]ethoxy}benzamide dihydrochloride
Synonyms:
2-hydroxy-5-{2-[(2-hydroxy-3-{4-[1-methyl-4-(trifluoromethyl)imidazol-2-yl]phenoxy}propyl)amino]ethoxy}benzamide dihydrochloride
Molecular Weight (g/mol):
567.39
Molecular Formula:
C23H27Cl2F3N4O5
InChi Key:
PURFQCFKYNMIQF-UHFFFAOYNA-N
SMILES:
Cl.Cl.CN1C=C(N=C1C1=CC=C(OCC(O)CNCCOC2=CC(C(N)=O)=C(O)C=C2)C=C1)C(F)(F)F
1
–
1
of
1
results
1
R&D Systems™ CGP 20712 dihydrochloride
CAS: 1216905-73-5 Molecular Formula: C23H27Cl2F3N4O5