CAS RN 1217674-10-6
IUPAC Name:
N,3-dimethyl-N-[4-(4-methylpiperidin-1-yl)butan-2-yl]benzene-1-sulfonamide hydrochloride
Synonyms:
N,3-dimethyl-N-[4-(4-methylpiperidin-1-yl)butan-2-yl]benzenesulfonamide hydrochloride
Molecular Weight (g/mol):
374.97
Molecular Formula:
C18H31ClN2O2S
InChi Key:
UIZKHTBWJSUGOV-UHFFFAOYNA-N
SMILES:
Cl.CC(CCN1CCC(C)CC1)N(C)S(=O)(=O)C1=CC=CC(C)=C1
1
–
1
of
1
results
1
Tocris Bioscience™ SB 258719 hydrochloride
CAS: 1217674-10-6 Molecular Formula: C18H31ClN2O2S