Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SB 258719 hydrochloride
SDP

Catalog No. 272650
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-265-0 50 mg
27-261-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 27-265-0 Supplier Tocris Bioscience™ Supplier No. 2726/50

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective 5-HT7 antagonist

Selective 5-HT7 receptor antagonist that displays > 100-fold selectivity over a range of other receptors. Reverses the hypothermic effect of 5-CT in mice following i.p. administration.

Chemical Identifiers

CAS 1217674-10-6
Molecular Formula C18H31ClN2O2S

Specifications

Quantity 50 mg
Formula Weight 374.97
Purity >99%
Molecular Weight (g/mol) 374.968
Chemical Name or Material SB 258719 hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.