accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 121788-77-0

IUPAC Name: N-[1,2-diphenyl-2-(trifluoromethanesulfonamido)ethyl]-1,1,1-trifluoromethanesulfonamide
Synonyms: N-[1,2-diphenyl-2-(trifluoromethanesulfonamido)ethyl]-1,1,1-trifluoromethanesulfonamide
Molecular Weight (g/mol): 476.41
Molecular Formula: C16H14F6N2O4S2
InChi Key: XQAIGOHPAZPGOU-UHFFFAOYNA-N
SMILES: FC(F)(F)S(=O)(=O)NC(C(NS(=O)(=O)C(F)(F)F)C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results