Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S,S)-N,N'-Bis(trifluoromethanesulfonyl)-1,2-diphenylethylenediamine, TCI America™
SDP

Catalog No. D25211G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25211G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D25211G Supplier TCI America Supplier No. D25211G

Chemical Identifiers

CAS 121788-77-0
Molecular Formula C16H14F6N2O4S2
Molecular Weight (g/mol) 476.41
MDL Number MFCD01321159
InChI Key XQAIGOHPAZPGOU-UHFFFAOYNA-N
Synonym (S,S)-1,2-Bis(trifluoromethanesulfonamido)-1,2-diphenylethane
PubChem CID 7009144
IUPAC Name N-[1,2-diphenyl-2-(trifluoromethanesulfonamido)ethyl]-1,1,1-trifluoromethanesulfonamide
SMILES FC(F)(F)S(=O)(=O)NC(C(NS(=O)(=O)C(F)(F)F)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 1 g
Formula Weight 476.41
Chemical Name or Material (S,S)-N,N′-Bis(trifluoromethanesulfonyl)-1,2-diphenylethylenediamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.