accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CFNOO

CAS RN 1218791-09-3

IUPAC Name: 2-(4-fluoro-3-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(4-fluoro-3-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 267.06
Molecular Formula: C12H15BFNO4
InChi Key: QBMHYMHOXGOPAF-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(=C(F)C=C1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results