CAS RN 1219168-18-9
IUPAC Name:
4-(6-{4-[2-(piperidin-1-yl)ethoxy]phenyl}pyrazolo[1,5-a]pyrimidin-3-yl)pyridine dihydrochloride
Synonyms:
4-(6-{4-[2-(piperidin-1-yl)ethoxy]phenyl}pyrazolo[1,5-a]pyrimidin-3-yl)pyridine dihydrochloride
Molecular Weight (g/mol):
472.41
Molecular Formula:
C24H27Cl2N5O
InChi Key:
RJDVIJJQKMGPMV-UHFFFAOYSA-N
SMILES:
Cl.Cl.C(CN1CCCCC1)OC1=CC=C(C=C1)C1=CN2N=CC(=C2N=C1)C1=CC=NC=C1
1
–
1
of
1
results
1
R&D Systems™ Dorsomorphin dihydrochloride
CAS: 1219168-18-9 Molecular Formula: C24H27Cl2N5O