accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOOCl

CAS RN 122-88-3

IUPAC Name: 2-(4-chlorophenoxy)acetic acid
Synonyms: sure-set caswell no. 204 4-chlorphenoxyessigsaeure 4CPA tomatotone PCPA parachlorophenoxyacetic acid marks 4-cpa tomato hold tomato fix
Molecular Weight (g/mol): 186.59
Molecular Formula: C8H7ClO3
InChi Key: SODPIMGUZLOIPE-UHFFFAOYSA-N
SMILES: OC(=O)COC1=CC=C(Cl)C=C1
Molecular Weight (g/mol) 
  • (4)
  • (5)
Percent Purity 
  • (3)
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (5)
  • (4)
  • (5)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)