accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 122030-04-0

IUPAC Name: 1,3,5-trifluoro-2-(trifluoromethyl)benzene
Synonyms: 1,3,5-trifluoro-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 200.08
Molecular Formula: C7H2F6
InChi Key: SGPXFVDJRSZIPK-UHFFFAOYSA-N
SMILES: FC1=CC(F)=C(C(F)=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results