CAS RN 122138-83-4
IUPAC Name:
7-fluoro-6-nitro-3,4-dihydro-2H-1,4-benzoxazine
Synonyms:
7-fluoro-6-nitro-3,4-dihydro-2H-1,4-benzoxazine
Molecular Weight (g/mol):
198.15
Molecular Formula:
C8H7FN2O3
InChi Key:
PEOVRVSERLAIQM-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=C(F)C=C2OCCNC2=C1
1
–
1
of
1
results
1
7-Fluoro-6-nitro-3,4-dihydro-2H-1,4-benzoxazine, 97%, Thermo Scientific Chemicals
CAS: 122138-83-4 Molecular Formula: C8H7FN2O3 Molecular Weight (g/mol): 198.153 MDL Number: MFCD23379858 InChI Key: PEOVRVSERLAIQM-UHFFFAOYSA-N PubChem CID: 14422956 IUPAC Name: 7-fluoro-6-nitro-3,4-dihydro-2H-1,4-benzoxazine SMILES: C1COC2=CC(=C(C=C2N1)[N+](=O)[O-])F