accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12225-21-7

AlHOONNONNSOOOSOHOOHOONNONNSOOOSOHOOHOONNONNSOOOSOHOO(E/Z)(E/Z)(E/Z)

CAS RN 12225-21-7

IUPAC Name: aluminium(3+) tris(4-{2-[3-carboxy-5-oxo-1-(4-sulfophenyl)-4,5-dihydro-1H-pyrazol-4-yl]diazen-1-yl}benzene-1-sulfonate)
Synonyms: acid yellow 23 aluminum lake
Molecular Weight (g/mol): 1429.19
Molecular Formula: C48H33AlN12O27S6
InChi Key: WNVDZDLVUGYYKK-UHFFFAOYNA-K
SMILES: [Al+3].OC(=O)C1=NN(C(=O)C1N=NC1=CC=C(C=C1)S([O-])(=O)=O)C1=CC=C(C=C1)S(O)(=O)=O.OC(=O)C1=NN(C(=O)C1N=NC1=CC=C(C=C1)S([O-])(=O)=O)C1=CC=C(C=C1)S(O)(=O)=O.OC(=O)C1=NN(C(=O)C1N=NC1=CC=C(C=C1)S([O-])(=O)=O)C1=CC=C(C=C1)S(O)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results