accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12228-87-4

IUPAC Name: 4-[(4-hydroxy-1,3,2,4-dioxadiboretan-2-yl)oxy]-1,3,2,4-dioxadiboretan-2-ol diamine tetrahydrate
Synonyms: 4-[(4-hydroxy-1,3,2,4-dioxadiboretan-2-yl)oxy]-1,3,2,4-dioxadiboretan-2-ol diamine tetrahydrate
Molecular Weight (g/mol): 263.37
Molecular Formula: B4H16N2O11
InChi Key: QZNWNKFWDJRMLV-UHFFFAOYSA-N
SMILES: N.N.O.O.O.O.OB1OB(OB2OB(O)O2)O1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)