Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boron 10 Isotope Standard For ICP/MS, SPEX CertiPrep™
SDP

Catalog No. ISOTB10 Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
125 mL
Packaging:
Triple-leached LDPE bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
ISOTB10 125 mL Triple-leached LDPE bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ISOTB10 Supplier SPEX CertiPrep Supplier No. ISOTB10
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
B1010μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 12228-87-4 , 7732-18-5
Molecular Formula B4H16N2O11
Molecular Weight (g/mol) 263.371
InChI Key QZNWNKFWDJRMLV-UHFFFAOYSA-N
Synonym ammonium tetraborate tetrahydrate, 4-4-hydroxy-1,3,2,4-dioxadiboretan-2-yl oxy-1,3,2,4-dioxadiboretan-2-ol diamine tetrahydrate, ammonium tetraborate eae r masculineiii, ammonium biborate tetrahydrate b2o3 basis, 4,4'-oxydi 1,3,2,4-dioxadiboretan-2-ol-ammonia-water 1/2/4
PubChem CID 44134537
IUPAC Name azane;2-hydroxy-4-[(4-hydroxy-1,3,2,4-dioxadiboretan-2-yl)oxy]-1,3,2,4-dioxadiboretane;tetrahydrate
SMILES B1(OB(O1)OB2OB(O2)O)O.N.N.O.O.O.O

Specifications

CAS Max % 99.99, 0.0
Physical Form Liquid
Quantity 125 mL
Grade Claritas PPT
Concentration or Composition (by Analyte or Components) 10mg/L
Packaging Triple-leached LDPE bottle
Chemical Name or Material Boron 10 Isotope Standard, For ICP/MS
Hazard Category Causes skin irritation, serious eye irritation
DOTInformation Hazardous
Recommended Storage Ambient

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.