accessibility menu, dialog, popup

UserName

Viewing profile as user:
CrAs

CAS RN 12254-85-2

IUPAC Name: chromium(3+) arsanetriide
Synonyms: chromium(3+) arsanetriide
Molecular Weight (g/mol): 126.92
Molecular Formula: AsCr
InChi Key: OAMWSOUCVTVANH-UHFFFAOYSA-N
SMILES: [Cr+3].[As-3]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results