CAS RN 1226917-67-4
IUPAC Name:
N-{4-[2-(6-cyano-1,2,3,4-tetrahydroisoquinolin-2-yl)ethyl]cyclohexyl}quinoline-4-carboxamide dihydrochloride
Synonyms:
N-{4-[2-(6-cyano-3,4-dihydro-1H-isoquinolin-2-yl)ethyl]cyclohexyl}quinoline-4-carboxamide dihydrochloride
Molecular Weight (g/mol):
511.49
Molecular Formula:
C28H32Cl2N4O
InChi Key:
HEZIOTGUXSPDAK-UHFFFAOYSA-N
SMILES:
Cl.Cl.O=C(NC1CCC(CCN2CCC3=C(C2)C=CC(=C3)C#N)CC1)C1=C2C=CC=CC2=NC=C1
1
–
1
of
1
results
1
R&D Systems™ SB 277011A dihydrochloride
CAS: 1226917-67-4 Molecular Formula: C28H32Cl2N4O