accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHOClHBr

CAS RN 1227675-51-5

IUPAC Name: 5-chloro-10-methyl-10-azatetracyclo[9.8.0.0²,⁷.0¹⁴,¹⁹]nonadeca-2,4,6,14,16,18-hexaen-4-ol hydrobromide
Synonyms: 5-chloro-10-methyl-10-azatetracyclo[9.8.0.0²,⁷.0¹⁴,¹⁹]nonadeca-2,4,6,14,16,18-hexaen-4-ol hydrobromide
Molecular Weight (g/mol): 394.74
Molecular Formula: C19H21BrClNO
InChi Key: GAUWIDFICGEZKR-UHFFFAOYNA-N
SMILES: Br.CN1CCC2=CC(Cl)=C(O)C=C2C2C1CCC1=CC=CC=C21

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)