accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNNNHNH2NNHH3COHN

CAS RN 1227911-45-6

IUPAC Name: 1-[6-(3-amino-1H-indazol-6-yl)-2-(methylamino)pyrimidin-4-yl]-N-cyclohexyl-6-methylpiperidine-3-carboxamide
Synonyms: 1-[6-(3-amino-1H-indazol-6-yl)-2-(methylamino)pyrimidin-4-yl]-N-cyclohexyl-6-methylpiperidine-3-carboxamide
Molecular Weight (g/mol): 462.6
Molecular Formula: C25H34N8O
InChi Key: QLPHOXTXAKOFMU-UHFFFAOYNA-N
SMILES: CNC1=NC(=CC(=N1)N1CC(CCC1C)C(=O)NC1CCCCC1)C1=CC=C2C(N)=NNC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)