CAS RN 122955-13-9
IUPAC Name:
10,10-bis[(pyridin-4-yl)methyl]-9,10-dihydroanthracen-9-one dihydrochloride
Synonyms:
10,10-bis(pyridin-4-ylmethyl)anthracen-9-one dihydrochloride
Molecular Weight (g/mol):
449.38
Molecular Formula:
C26H22Cl2N2O
InChi Key:
WOGWMARIFDNZON-UHFFFAOYSA-N
SMILES:
Cl.Cl.O=C1C2=CC=CC=C2C(CC2=CC=NC=C2)(CC2=CC=NC=C2)C2=CC=CC=C12
1
–
1
of
1
results
1
R&D Systems™ XE 991 dihydrochloride
CAS: 122955-13-9 Molecular Formula: C26H22Cl2N2O