accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOONOSFHOOH(E/Z)

CAS RN 1229652-21-4

IUPAC Name: (3-{[4-({3-[(4-fluorophenyl)methyl]-2,4-dioxo-1,3-thiazolidin-5-ylidene}methyl)phenoxy]methyl}phenyl)boronic acid
Synonyms: 3-[4-({3-[(4-fluorophenyl)methyl]-2,4-dioxo-1,3-thiazolidin-5-ylidene}methyl)phenoxymethyl]phenylboronic acid
Molecular Weight (g/mol): 463.29
Molecular Formula: C24H19BFNO5S
InChi Key: VTNKMYWFWQTEHE-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(COC2=CC=C(C=C3SC(=O)N(CC4=CC=C(F)C=C4)C3=O)C=C2)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results