accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 123-04-6

IUPAC Name: 3-(chloromethyl)heptane
Synonyms: 3-(chloromethyl)heptane
Molecular Weight (g/mol): 148.67
Molecular Formula: C8H17Cl
InChi Key: WLVCBAMXYMWGLJ-UHFFFAOYNA-N
SMILES: CCCCC(CC)CCl
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)