Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(chloromethyl)heptane, 90%, Thermo Scientific™

Catalog No. AC344981000
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC344981000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC344981000 Supplier Thermo Scientific Chemicals Supplier No. 344981000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-04-6
Molecular Formula C8H17Cl
Molecular Weight (g/mol) 148.674
MDL Number MFCD00018940
InChI Key WLVCBAMXYMWGLJ-UHFFFAOYSA-N
Synonym 3-chloromethyl heptane, 2-ethylhexyl chloride, 1-chloro-2-ethylhexane, heptane, 3-chloromethyl, hexane, 1-chloro-2-ethyl, 2-ethylhexylchlorid, 2-ethylhexylchloride, acmc-209any, 4-01-00-00430 beilstein handbook reference
PubChem CID 31240
IUPAC Name 3-(chloromethyl)heptane
SMILES CCCCC(CC)CCl

Specifications

CAS Min % 88.0
CAS Max % 100.0
Color Yellow
Density 0.8820g/mL
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Refractive Index 1.4320 to 1.4360 (20°C, 589nm)
Linear Formula H3C(CH2)3CH(CH2Cl)CH2CH3
Quantity 100 mL
Formula Weight 148.68
Percent Purity ≥88%
Physical Form Liquid
Chemical Name or Material 3-(Chloromethyl)heptane
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.