Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Chloromethyl)heptane 98.0+%, TCI America™
SDP

Catalog No. C15345G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C15345G Supplier TCI America Supplier No. C15345G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 123-04-6
Molecular Formula C8H17Cl
Molecular Weight (g/mol) 148.674
MDL Number MFCD00018940
InChI Key WLVCBAMXYMWGLJ-UHFFFAOYSA-N
Synonym 3-chloromethyl heptane, 2-ethylhexyl chloride, 1-chloro-2-ethylhexane, heptane, 3-chloromethyl, hexane, 1-chloro-2-ethyl, 2-ethylhexylchlorid, 2-ethylhexylchloride, acmc-209any, 4-01-00-00430 beilstein handbook reference
PubChem CID 31240
IUPAC Name 3-(chloromethyl)heptane
SMILES CCCCC(CC)CCl

Specifications

Color Colorless
Boiling Point 169°C
Quantity 5 g
UN Number 1993
Formula Weight 148.67
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(Chloromethyl)heptane
EINECSNumber (2)-0066
RTECSNumber MI8760000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.