accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH2

CAS RN 123-20-6

IUPAC Name: ethenyl butanoate
Synonyms: vinyl butyrate
Molecular Weight (g/mol): 114.14
Molecular Formula: C6H10O2
InChi Key: MEGHWIAOTJPCHQ-UHFFFAOYSA-N
SMILES: CCCC(=O)OC=C
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results