accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 123-95-5

IUPAC Name: butyl octadecanoate
Synonyms: butyl stearate N-butyl stearate
Molecular Weight (g/mol): 340.59
Molecular Formula: C22H44O2
InChi Key: ULBTUVJTXULMLP-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCC(=O)OCCCC
Molecular Weight (g/mol) 
  • (1)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (5)
  • (1)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)