Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyl Stearate, Mixture of ca 55-65% Stearyl- and 25-40% Cetyl-chain, Thermo Scientific™

Catalog No. AC403685000
Change view
Click to view available options
Quantity:
500 g
3 kg
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC403685000 500 g
AC403680200 20 L
AC403680030 3 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC403685000 Supplier Thermo Scientific Chemicals Supplier No. 403685000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-95-5
Molecular Formula C22H44O2
Molecular Weight (g/mol) 340.59
MDL Number MFCD00026669
InChI Key ULBTUVJTXULMLP-UHFFFAOYSA-N
Synonym butyl stearate, n-butyl stearate, octadecanoic acid, butyl ester, kesscoflex bs, n-butyl octadecanoate, stearic acid, butyl ester, butyl octadecylate, kessco bsc, stearic acid n-butyl ester, polycizer 332
PubChem CID 31278
ChEBI CHEBI:85983
IUPAC Name butyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OCCCC

Specifications

Linear Formula CH3(CH2)16COO(CH2)3CH3
Quantity 500 g
Merck Index 15, 1592
Formula Weight 340.58
Chemical Name or Material Butyl stearate, mixture of ca. 55-65% stearyl- and 25-40% cetyl-chain
EINECSNumber 204-666-5
RTECSNumber WI2900000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.