accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2SOOONa

CAS RN 123333-70-0

IUPAC Name: sodium 4-aminobenzene-1-sulfonate
Synonyms: sodium sulfanilate
Molecular Weight (g/mol): 195.17
Molecular Formula: C6H6NNaO3S
InChi Key: KSVSZLXDULFGDQ-UHFFFAOYSA-M
SMILES: [Na+].NC1=CC=C(C=C1)S([O-])(=O)=O
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)