Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Sulfanilate Hydrate 98.0+%, TCI America™
SDP

Catalog No. S08325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S08325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S08325G Supplier TCI America Supplier No. S08325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 123333-70-0
Molecular Formula C6H6NNaO3S
Molecular Weight (g/mol) 195.17
MDL Number MFCD00151043
InChI Key KSVSZLXDULFGDQ-UHFFFAOYSA-M
Synonym sodium sulfanilate hydrate, sodium 4-aminobenzenesulfonate hydrate, sulfanilic acid sodium salt hydrate, 4-aminobenzenesulfonic acid, sodium salt hydrate, sodium 4-aminobenzenesulfonate hydrate 1:1:x, acmc-209aoo, c6h6nso3.na.h2o, sodium4-aminobenzenesulfonatehydrate, p-anilinesulfonic acid sodium
PubChem CID 23672401
IUPAC Name sodium 4-aminobenzene-1-sulfonate
SMILES [Na+].NC1=CC=C(C=C1)S([O-])(=O)=O

Specifications

Color White
Quantity 5 g
Formula Weight 195.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Sulfanilate Hydrate [for Biochemical Research]
EINECSNumber (3)-1972
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.