accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONOHNHNNNONCH3CH3

CAS RN 1234480-50-2

IUPAC Name: 5-{[2-ethoxy-4-(4-hydroxypiperidin-1-yl)phenyl]amino}-2,9-dimethyl-2,4,6,9-tetraazatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-10-one
Synonyms: 5-{[2-ethoxy-4-(4-hydroxypiperidin-1-yl)phenyl]amino}-2,9-dimethyl-2,4,6,9-tetraazatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-10-one
Molecular Weight (g/mol): 474.57
Molecular Formula: C26H30N6O3
InChi Key: QAPAJIZPZGWAND-UHFFFAOYSA-N
SMILES: CCOC1=C(NC2=NC=C3N(C)C(=O)C4=CC=CC=C4N(C)C3=N2)C=CC(=C1)N1CCC(O)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)