accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 123520-73-0

IUPAC Name: sodium 2-bromo-4-[(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)amino]benzen-1-olate
Synonyms: sodium 2-bromo-4-[(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)amino]benzenolate
Molecular Weight (g/mol): 457.88
Molecular Formula: C12H5Br3NNaO2
InChi Key: HNPUZWCYQDQTKJ-UHFFFAOYSA-M
SMILES: [Na+].[O-]C1=C(Br)C=C(C=C1)N=C1C=C(Br)C(=O)C(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results