accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1236858-52-8

IUPAC Name: 1-(oxan-4-yl)-6-[1-(3-phenoxyazetidin-1-yl)ethyl]-1H,2H,4H-pyrazolo[3,4-d]pyrimidin-4-one
Synonyms: 1-(oxan-4-yl)-6-[1-(3-phenoxyazetidin-1-yl)ethyl]-2H-pyrazolo[3,4-d]pyrimidin-4-one
Molecular Weight (g/mol): 395.46
Molecular Formula: C21H25N5O3
InChi Key: WEDCMPMDOVEMSV-UHFFFAOYNA-N
SMILES: CC(N1CC(C1)OC1=CC=CC=C1)C1=NC(=O)C2=CNN(C3CCOCC3)C2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results